KIMBLE® HI-VAC® Straight Valve, with PTFE Plug, without Tip O-Ring, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
826600-0004 826600-0008 826600-0012
KT826600-0004EA 97.92 CAD
KT826600-0004 KT826600-0008 KT826600-0012
KIMBLE® HI-VAC® Straight Valve, with PTFE Plug, without Tip O-Ring, DWK Life Sciences
Valves Vacuum Valves
The straight high-vacuum valve without tip O-ring is for use in vacuum applications up to 5×10-7torr.
  • Designed with a precision external thread for fine control
  • Valve can be used as a bleed up to ¹/₂ turn
  • Full open is at 1¹/₂ turns
Primary seal is made with a FKM O-ring on a tapered seat. Non-rotating PTFE shaft can be used continuously at system temperatures of 230 °C or intermittently at 275 °C. Manufactured from 33 expansions, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.