CryoCube® F570h, ULT Upright Freezers, Eppendorf
Supplier: Eppendorf
F571242035 F571243035
75874-440EA 40080.52 CAD
75874-440 75874-442 75874-718 75874-720 75874-722 CA76534-696 CA76534-698 MPCA76534-692
CryoCube® F570h, ULT Upright Freezers, Eppendorf
Freezers Ultra Low Temperature Freezers
The Eppendorf ULT Upright Freezers are known for their high energy efficiency and they combine all the enhanced technology with additional vacuum insulation coverage.
  • Reduced energy consumption with high-efficiency fan, compressor and condenser
  • Easy opening with new ergonomic handle that requires less force
  • Easy access to samples with new magnetic closures on inner doors
  • Storage up to 40000 vessels
The ultra-low temperature freezers combine maximum sample security with improved functionality. The advancements decrease power consumption and make freezers more comfortable to use.