Disposable Cuvettes for Fluorometer, FireflySci
Supplier: FIREFLYSCI, INC.
1FLPMMA 1FLPS 9FLPMMA
76016-580EA 239.37 CAD
76016-580 76016-582 76016-592
Disposable Cuvettes for Fluorometer, FireflySci
Cuvettes Spectrophotometer Cuvettes
These cuvettes are used for rapid assays and comes in different materials polystyrene and acrylic.
  • Four windows and base optically polished
  • With open top