Measuring Cylinders with Hexagonal Base, United Scientific Supplies
Supplier: United Scientific Supplies
80031 80032 80033 80034 80035 80036 80037 80038
76263-182PK 100.3 CAD
76263-182 76263-184 76263-186 76263-188 76263-190 76263-192 76263-194 470339-366
Measuring Cylinders with Hexagonal Base, United Scientific Supplies
Cylinders Measuring Cylinders
Unbreakable polypropylene construction.
  • Stable hexagonal base
  • Unbreakable Polypropylene
Polypropylene one-piece measuring cylinders have raised graduations for easy readability. They are autoclavable and have excellent contact clarity. The hexagonal base provides excellent stability, and the tapered spout enables easy pouring of reagents. Stated tolerances are the same as graduated intervals below.