Griffin Style Beakers, United Scientific Supplies
Supplier: United Scientific Supplies
BP0050 BP0100 BP0250-VWR BP0500 BP0600-VWR BP1000-VWR BP2000-VWR
76263-370PK 35.31 CAD
76263-370 76263-372 76263-374 76263-376 76263-378 76263-380 76263-382
Griffin Style Beakers, United Scientific Supplies
Beakers
Autoclavable polypropylene nesting beakers.
  • Raised graduations
  • Autoclavable
Beakers, molded in polypropylene, have excellent clarity and very good chemical resistance. They are autoclavable and feature raised graduations on the outer surface. Griffin Style Beakers nest without sticking together.