Siphon Pumps, National Spencer
Supplier: NATIONAL SPENCER, INC. MS
10010 10020
MSPP-10010BX 663.08 CAD
MSPP-10010 MSPP-10020
Siphon Pumps, National Spencer
Pumps Drum/Transfer Pumps
Siphon pump made of polyethylene body and polypropylene suction tube.
  • Two years limited warranty
  • Compatible with alcohol, anti-freeze, water-based and windshield washer fluids
Ordering information: Small pump must be ordered in the multiples of 120 and large pump in the multiples of 24.