2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2,2'-Azobis[2-(2-imidazolin-2-yl)propane] Dihydrochloride, 1,2-Bis(2-(4,5-dihydro-1H-imidazol-2-yl)propan-2-yl)diazene dihydrochloride, 2-{2-[(E)-2-[2-(4,5-Dihydro-1H-imidazol-2-yl)proan-2-yl]diazen-1-yl]propan-2-yl}-4,5-dihydro-1H-imidazole dihydrochloride

A3012-25G A3012-100G
76218-780EA 58.87 CAD
76218-780 76219-282
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride ≥98.0% (by HPLC)
2,2'-Azobis[2-(2-imidazolin-2-yl)propane] dihydrochloride
Formula: C₁₂H₂₄Cl₂N₆
MW: 323.27 g/mol
MDL Number: MFCD00142723
CAS Number: 27776-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83685ce18ea8dbef810a5f4e1363d441cd82f7ad&vid=83685ce18ea8dbef810a5f4e1363d441cd82f7ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) Min. 98.0 area%
Elemental Analysis (Nitrogen) 25.00 - 27.00%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR