Tubing Connectors, CADENCE SCIENCE®
Supplier: Cadence Science
6184IND 6217IND 6151IND 6153IND 6158IND 6219IND 6220IND 6411IND 6413IND 6453IND 6512IND 6515IND 6159IND
76268-958EA 139.86 CAD
76268-958 76268-964 76268-990 76268-992 76268-994 76268-996 76268-998 76269-000 76269-002 76269-004 76269-016 76269-018 76269-020 76269-022 76269-034 76269-048 76269-050 76269-052 76269-054 76269-056 76269-058 76269-104
Tubing Connectors, CADENCE SCIENCE®
Fittings and Connectors
These tubing connectors are used in hospitals, laboratories, and biomedical instruments. The metal connectors are precision machined and are made from either nickel plated brass or 316 stainless steel.