Laboratory Scoops, United Scientific Supplies
Supplier: United Scientific Supplies
81260 81253 81254 81255 81256 81257 81258 81259
76263-062EA 49.07 CAD
76263-062 76263-288 76263-290 76263-292 76263-294 76263-296 76263-298 76263-300
Laboratory Scoops, United Scientific Supplies
Scoops
Conveniently designed handled scoops.
  • Autoclavable
These sturdy polypropylene scoops are available in a variety of sizes, as well as in a complete set of seven scoops. Capacities are molded into the top of each scoop for easy reference. The polypropylene material is autoclavable and will not rust or corrode.