Buchner Funnels, United Scientific Supplies
Supplier: United Scientific Supplies
57105-VWR 57105-A-PK/12 57106-PK/12
76263-244PK 183.96 CAD
76263-244 76263-246 76263-248
Buchner Funnels, United Scientific Supplies
Funnels Filtration Funnels Büchner Funnels
The top portion of these polypropylene, two-piece funnels are snaps into the lower one, forming a vacuum seal that does not require greasing. The base of the top portion has a perforated filter plate.
  • Lightweight and easy to clean
  • Autoclavable
Useable with or without filter paper.