Powder Funnels, PP, United Scientific Supplies
Supplier: United Scientific Supplies
57221 57222 57223 57224
76263-230PK 14.72 CAD
76263-230 76263-232 76263-234 76263-236
Powder Funnels, PP, United Scientific Supplies
Funnels Powder Funnels
Useful for adding powder chemical to reaction vessels.
  • Autoclavable
Polypropylene funnels have short, wide stems for convenient use with powders.