Utility Funnels, United Scientific Supplies
Supplier: United Scientific Supplies
FPPC35 FPPC52 FPPC65 FPPC03-VWR
76263-222EA 13.95 CAD
76263-222 76263-224 76263-226 76263-228
Utility Funnels, United Scientific Supplies
Funnels Powder Funnels
Useful for adding powder chemical to reaction vessels.
  • Polypropylene
  • Autoclavable
These funnels have eight internal ribs to facilitate filtration. They are made of polypropylene.
Order Now
SPECIFICATIONS
Environmentally Preferable