Funnels with Standard Stems, United Scientific Supplies
Supplier: United Scientific Supplies
FSPP55-VWR FSPP65-VWR FSPP75-VWR FSPP10-VWR FSPP150
76263-206PK 12.15 CAD
76263-206 76263-208 76263-210 76263-212 76263-214
Funnels with Standard Stems, United Scientific Supplies
Funnels Filtration Funnels Filter Funnels
These clear funnels have long stems, smooth walls, and external ribs to prevent air lock.
  • Autoclavable
  • Molded in polypropylene
These long stemmed funnels allow you to fill tall bottles.