Carboys, United Scientific Supplies
Supplier: United Scientific Supplies
34001 34002
76263-154EA 95.79 CAD
76263-154 76263-156
Carboys, United Scientific Supplies
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
  • Autoclavable
  • Wide handles for convenient carrying and pouring
These polypropylene carboys are ideal for storing various laboratory solutions. Threads should be completely disengaged before autoclaving. Mouth Int.Ø is 67 mm.