Wash Bottle, Unitary, United Scientific Supplies
Supplier: United Scientific Supplies
36607 36605 36606
76263-144PK 83.27 CAD
76263-144 76263-146 76263-148
Wash Bottle, Unitary, United Scientific Supplies
Bottles Wash Bottles
Low density polyethylene wash bottles enable efficient laboratory work.
  • Built-in delivery tube
  • Translucent
  • Raised graduations
The distinguishing feature of these bottles is the built-in delivery tube which makes it a one-piece wash bottle. Graduated on the outer surface to reflect an approximate volume of solution filled or dispensed.