UniStore™ Reagent Bottles, Narrow Mouth, HDPE
Supplier: United Scientific Supplies
33401 33402 33403 33404 33405
76263-120PK 35.09 CAD
76263-120 76263-122 76263-124 76263-126 76263-128
UniStore™ Reagent Bottles, Narrow Mouth, HDPE
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Translucent
  • Pliable
UniStore™ narrow mouth HDPE reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.