UniStore™ Reagent Bottles, Wide Mouth, PP
Supplier: United Scientific Supplies
33311 33306 33307 33308 33309 33310
76263-108PK 18.94 CAD
76263-108 76263-110 76263-112 76263-114 76263-116 76263-118
UniStore™ Reagent Bottles, Wide Mouth, PP
Bottles Plastic Bottles
Bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Translucent
  • Rigid
  • Autoclavable
UniStore™ wide mouth PP reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant. Excellent chemical resistance to most acids, bases, and alcohols.
Ordering information: Polypropylene caps are included.