Unisette™ Biopsy Processing Cassette in Quickload™ Stacks for Primera Printers, Simport Scientific
Supplier: Simport Scientific
M406-12T M406-2T M406-3T M406-4T M406-5T M406-6T M406-7T M406-8T M406-9T M406-10T M406-11T
CA76214-030CS 349.8 CAD
CA76214-030 CA76214-348 CA76214-350 CA76214-352 CA76214-354 CA76214-356 CA76214-358 CA76214-360 CA76214-362 CA76214-364 CA76214-366
Unisette™ Biopsy Processing Cassette in Quickload™ Stacks for Primera Printers, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
Made of acetal and resistant to the chemical action of most histological solvents and suitable for automated printers.
  • In Quickload™ stacks, for Primera printers
  • 35° anterior writing area
  • One-piece construction with hinge-lock design prevents early separation of base and lid
  • Snap latch and hinge-lock design; cassettes will always relock securely
  • Allows for one-hand operation
Secure specimens during processing with the Unisette™ Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design allows for one-hand operation and always relocks securely without danger of specimen loss.
Certifications: CE-IVD.
Packaging: Each case contains 25 Quickload™ stacks of 40 cassettes.