Slimsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Supplier: Simport Scientific
M510-2T M510-3T M510-12T M510-4T M510-5T M510-6T M510-7T M510-8T M510-9T M510-10T M510-11T
CA76214-144CS 1363.26 CAD
CA76214-144 CA76214-146 CA76214-148 CA76214-240 CA76214-242 CA76214-244 CA76214-246 CA76214-248 CA76214-250 CA76214-252 CA76214-254
Slimsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
Secure specimens during processing with the Slimsette™ Biopsy Processing/Embedding Cassette, in Quickload™ stacks.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvent
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes.