Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Supplier: Simport Scientific
M505-8T M505-9T M505-10T M505-11T M505-2T M505-3T M505-4T M505-5T M505-7T M505-12T M505-6T
CA76214-100CS 431.07 CAD
CA76214-100 CA76214-102 CA76214-104 CA76214-106 CA76214-192 CA76214-194 CA76214-196 CA76214-198 CA76214-202 CA95045-518 CA95045-526
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Embedding Cassettes Tissue Processing Cassettes
Secure specimens during processing with the Unisette™ Tissue Processing/Embedding Cassette.
  • In Quickload™ stacks
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevents early separation of base and lid
  • Snap latch and hinge-lock design; cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
The snap latch and hinge-lock design allows for one-hand operation and always relocks securely without danger ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable