O-Rings, Perfluoro, Chemglass
Supplier: Chemglass
CG-309-117 CG-309-136 CG-309-137 CG-309-205 CG-309-223 CG-309-331 CG-309-105 CG-309-218 CG-309-006 CG-309-007 CG-309-009 CG-309-010 CG-309-013 CG-309-014 CG-309-018 CG-309-022 CG-309-107 CG-309-108 CG-309-110 CG-309-114 CG-309-115 CG-309-116 CG-309-120 CG-309-121 CG-309-204 CG-309-208 CG-309-210 CG-309-212 CG-309-214 CG-309-217 CG-309-222 CG-309-226 CG-309-019 CG-309-024
89027-268EA 63.12 CAD
89027-268 89027-278 89027-292 89027-306 89027-308 89027-310 89027-316 89027-332 89027-338 89027-340 89027-344 89027-348 89029-318 89029-322 89029-324 89029-332 89027-248 89027-250 89027-252 89027-254 89027-256 89027-258 89027-260 89027-262 89027-264 89027-266 89027-270 89027-272 89027-274 89027-276 89027-280 89027-282 89027-284 89027-286 89027-288 89027-290 89027-296 89027-298 89027-300 89027-304 89027-312 89027-314 89027-318 89027-320 89027-322 89027-324 89027-326 89027-330 89027-334 89027-336 89027-342 89029-312 89029-314 89029-316
O-Rings, Perfluoro, Chemglass
Seals Sealing Rings
Perfluoro is a family of perfluoroelastomer compounds designed to provide superior chemical resistance, similar O-rings such as Kalrez® 4079 and Chemraz®.
  • Perfluoro O-rings are stocked in WHITE 75 durometer
  • Effective service temperature is –7 to 230 °C
  • Used with Duran flanges
These O-rings are used with acids, inorganic bases, alcohols/ethers, aromatic hydro-carbons, esters, aldehydes/ ketones, aliphatic hydrocarbons, amines and chlorocarbon solvents.