Nunc™ High Density Cell Factory™ Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
169102 169118 169160
10847-596CS 4309.79 CAD
10847-596 10847-598 10847-600
Nunc™ High Density Cell Factory™ Systems, Thermo Scientific
Bioreactors
High density cell factory systems provide a simple solution for increasing cell culture yields.
  • 30% more surface area and yield within the standard system footprint
  • Increase labor and handling efficiencies by achieving more output in a single process run
  • Increase manufacturing capacity
Its has additional layers. Cell factory systems are designed to optimize any process by increasing output, while maintaining the same manufacturing footprint as with the standard cell factory system.
Consume less and reduce decontamination and waste disposal costs.
Order Now
SPECIFICATIONS
Environmentally Preferable