ULTROL® Grade Zwitterionic Buffer, CAPS, Calbiochem®, MilliporeSigma
Supplier: MilliporeSigma
239782-100GM 239782-1KG
CA80057-968EA 372.86 CAD
CA80057-968 CA80057-970
ULTROL® Grade Zwitterionic Buffer, CAPS, Calbiochem®, MilliporeSigma
Buffers Biological Buffers
Zwitterionic buffer is useful in the pH 7.9 – 11.1 range and has a pKa of 10.0 – 10.5 at 25 °C.
Has an absorbance of ≤0.05 (100 mM, 260 nM).