BZ-Lys-OH ≥95%

Supplier: CHI Scientific

5-03019 5-03020
76224-119EA 206.65 CAD
76224-119 76224-121
BZ-Lys-OH ≥95%
BZ-Lys-OH

Caution: Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp

Formula: C₁₃H₁₈N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00038205
CAS Number: 366-74-5

Order Now

Specification Test Results

Protease-free Yes
Animal-free Yes
Endotoxin Level low (0.1 ng/μg)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR