Swingsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Supplier: Simport Scientific
M518-11T M518-12T M518-2T M518-3T M518-4T M518-5T M518-6T M518-7T M518-8T M518-9T M518-10T
CA76214-230CS 818.13 CAD
CA76214-230 CA76214-232 CA76214-256 CA76214-258 CA76214-260 CA76214-262 CA76214-264 CA76214-368 CA76214-370 CA76214-372 CA76214-374
Swingsette™ Biopsy Processing/Embedding Cassette in Quickload™ Stacks, Simport Scientific
Embedding Cassettes Biopsy Processing Cassettes
Secure specimens during processing with the Swingsette™ Biopsy Processing/Embedding Cassette, in Quickload™ stacks.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Easy to assemble hinged lids
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.