2-Propanol solution 70% in water ACS, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
LP420-204P
LP420-4P
LP420-5G
BJLP420-204PEA
5428.3
CAD
BJLP420-204P
BJLP420-4P
BJLP420-5G
2-Propanol solution 70% in water ACS, Burdick & Jackson™
2-Propanol
ACS general use certified, reagent grade
- Low residue
- Low water
- High GC purity
Formula:
(CH₃)₂CHOH Melting Pt: –62.1 °C Density: 0.785 g/cm³ (25 °C) (pure substance) Flash Pt: 11.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 UN: 1219 ADR: 3,II |
|
Specification Test Results
Appearance | Passes test |
Color (APHA) | 10 max. |
Isopropyl Alcohol (v/v) | 68 - 72 % |
Specific Gravity (20ºC/20ºC) | 0.863 - 0.883 |
Water (v/v) | 28 - 32 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...