Phenylarsine oxide solution_standard 0.00564 N
Supplier: Hach
Synonyms:
PAO, Oxophenylarsine, Arzene
199953
199917
CA11016-622EA
184.58
CAD
CA11016-622
CA11024-738
Phenylarsine oxide solution_standard 0.00564 N
Phenylarsine oxide
For chlorine determination by amperometric titration methods.
- Available in digital titrator cartridges
- Instrument: AutoCat™ 9000
- Method: Amperometric Titration
- Platform: Solution
- Range: 0.5 mg/ l and above Cl₂
Formula:
C₆H₅AsO MW: 168.02 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00001990 CAS Number: 637-03-6 |
Specification Test Results
Instrument | AutoCat™ 9000 |
Method | 8334 |
Method Name | Amperometric Titration |
Method Number 2 | 10025 |
Method Number 3 | 8168 |
Platform | Solution |
Range | 0.5 mg/ L and above Cl2 |
Range 2 | 0.0051 - 5.000 mg/L Cl2 |
Range 3 | 0.0012 - 5.000 mg/L Cl2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...