Select-a-Size DNA Clean and Concentrator, MagBeads, Zymo Research
Supplier: Zymo Research
D4085 D4084
76197-066EA 1955.39 CAD
76197-066 76197-140
Select-a-Size DNA Clean and Concentrator, MagBeads, Zymo Research
Beads Magnetic Beads
The select-a-size DNA MagBead kit provides the fastest and easiest method to purify specific ranges of DNA fragments from PCR, endonuclease digestions, ligations, library preparations, adapter removal, etc.
  • Size selection can be tuned from 100 bp to 1000 bp
  • Elution volume: ≥10 µl of DNA Elution Buffer
  • Eluted DNA is well suited for use in next generation sequencing, PCR, DNA ligation, endonuclease digestion, etc
This allows for specific cutoffs that can be modified to suit reaction clean-up, left-sided, right-sided or even double-sided size selection. The Select-a-Size MagBeads selectively binds fragments based on the volume of buffer added relative to the sample. Simply choose a desired cutoff to bind the target of interest onto the beads and remove species outside of this range. The desired DNA is easily eluted from the beads following a rapid wash regimen.
Size selections can be performed in as little as ten minutes to yield high-quality DNA which is suitable for highly sensitive applications including Next Generation Sequencing, and any other sensitive downstream applications. The protocol can be performed manually or using an automated platform for high throughput processing.