Vacuum Desiccator with White Base, United Scientific Supplies
Supplier: United Scientific Supplies
55204-VWR 55205 55206-VWR 55207
76263-196EA 145.11 CAD
76263-196 76263-198 76263-200 76263-202 470339-470 470339-892
Vacuum Desiccator with White Base, United Scientific Supplies
Desiccators Desiccators, Plastic
Compact design for easy use.
  • Autoclavable
  • Clear design
These autoclavable desiccators are made of polypropylene and polycarbonate, and can hold a vacuum up to 740 mm Hg for 24 hours without any greasing. Autoclavable at 121 °C for 15 minutes. The top dome, molded in rigid and transparent PC, gives a crystal-clear view of the sample. The knurled knob on the top provides easy handling of the dome.
The lower part is made of PP and includes a perforated PP desiccator plate. An internal groove on the flange holds the silicon rubber O-ring. The PPMore Product Information