Fine Mesh Basket, Sklar®
Supplier: Sklar
10-6604 10-6628 10-6620 10-6622 10-6630 10-6632 10-6634 10-6636 10-6729 10-6730 10-6732 10-6600 10-6602 10-6610 10-6612 10-6614 10-6616 10-6618 10-6606 10-6608 10-6624 10-6626
76191-492EA 1362.28 CAD
76191-492 76191-514 76191-584 76191-586 76191-692 76191-694 76191-696 76191-698 76191-700 76191-702 76191-704 76191-718 76191-720 76191-722 76191-842 76191-844 76191-846 76191-848 76193-030 76193-032 76193-038 76193-040
Fine Mesh Basket, Sklar®
Baskets Wire Baskets
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Fine Mesh Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, with or without removable hinges and with or without locks.