Ramelet Phlebectomy Hook, Sklar®
Supplier: Sklar
06-4174 06-4170 06-4173 06-4175 06-4172 06-4171
76191-524EA 747.71 CAD
76191-524 76191-526 76191-528 76191-482 76191-484 76193-036
Ramelet Phlebectomy Hook, Sklar®
Retractors
Sklar's® Ramelet Phlebectomy Hook is used to grip the vein during surgical procedures.
  • With industrial standard
  • With five major surgical instruments line
  • Delivers superior precision for any surgical procedure
The hook comes in fine and medium tips, angled left and right. The handle is round and knurled, which makes it easier to roll between the fingers for better control.