Sodium linoleate ≥99.0%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

02199875.1 0219987501 0219987505
76203-058EA 100.68 CAD
76203-058 76203-060 76203-062
Sodium linoleate ≥99.0%, white powder
Sodium linoleate

Linoleic acid (LA) is an unsaturated n-6 fatty acid; it belongs to one of the two families of essential fatty acids. The body cannot synthesize linoleic acid from other food components.


  • Purity : ≥99.0%
  • Key Applications : Antioxidant
  • Product Type : Biochemicals
  • Biochemical : Category Lipids
  • Presentation : White Powder
  • Format : Powder
  • Solubility : Solubility in methanol (50mg/mL - Clear, Colorless Solution.


Storage: under nitrogen.

Formula: C₁₈H₃₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00063202
CAS Number: 822-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd5cf184acf09841477a15bfb2376a71e84af8d9&vid=dd5cf184acf09841477a15bfb2376a71e84af8d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity 99.0%
FTIR Conforms to Standard C
Solubility (50mg/ml in methanol) Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR