2'-Deoxyadenosine monohydrate

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0219465405 0219465425
IC0219465405EA 335.48 CAD
IC0219465405 IC0219465425
2'-Deoxyadenosine monohydrate
2'-Deoxyadenosine monohydrate

Cell Culture ReagentWhite crystalline powderPurity: 98-100%Hydrate


  • Product Type : Biochemicals
  • Chemical Class : Nucleotides & Nucleic Acids

Formula: C₁₀H₁₃N₅O₃·1H₂O
Melting Pt: 186...196 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149364
CAS Number: 16373-93-6

Order Now

Specification Test Results

Identity Test Passes
Purity 98%
FTIR Conforms to Standard
Melting Point 186 - 196°C
UV/Vis: l max (water) OD250/OD260 OD280/OD260 259 ± 2 nm 0.70 - 0.85 0.10 - 0.16


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR