CleanMax® Clean and Sterile Disposable Coveralls with Elastic Wrist/Ankle, Lakeland Industries
Supplier: Lakeland
CTL417CS-MD CTL417CS-LG CTL417CS-XL CTL417CS-2X CTL417CS-3X CTL417CS-4X CTL417CS-5X
76193-328CS 1504.79 CAD
76193-328 76193-330 76193-332 76193-334 76193-336 76193-338 76193-340
CleanMax® Clean and Sterile Disposable Coveralls with Elastic Wrist/Ankle, Lakeland Industries
Disposable Garments
Clean and sterile disposable coveralls are individually double-bagged and folded to promote easier gowning with less excursions.
  • Clean and sterile
  • Manufactured in silicone-free environment
  • Latex-free
  • Tunneled elastic on wrists (with thumb loops), ankles, and back half of waist
  • Individually double-bagged
  • Certificate of radiation included
  • Gamma radiation indicator dots on each package
CleanMax® cleanroom garments provide comfort, quality, and protection. CleanMax® sterilized garments are both clean manufactured and sterile (not all sterile garments areMore Product Information