Anti-IgG Goat Antibody (Horseradish Peroxidase (HRP))

Supplier: MP Biomedicals
0855455
IC0855455EA 416.12 CAD
IC0855455
Anti-IgG Goat Antibody (Horseradish Peroxidase (HRP))
Antibodies
Product is the lyophilized powder of goat IgG fraction to mouse IgG (whole molecule) and buffer salts.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Horseradish Peroxidase (HRP)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR