DURAN® YOUTILITY® GL 45 Laboratory Bottle System, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
218812854 218813653 218814452 218815457 218862859 218863658 218864457 218865453
76264-182CS 99.82 CAD
76264-182 76264-184 76264-186 76264-188 76264-190 76264-192 76264-194 76264-196
DURAN® YOUTILITY® GL 45 Laboratory Bottle System, DWK Life Sciences
Bottles Glass Bottles
The new generation of glass laboratory bottles.
  • Borosilicate 3.3 pharmacopeia type 1 neutral glass bottles
  • Bottle tags are made of silicone and available in eight colors
  • Labels accept copier, printer, ballpoint, and technical markers and inks
  • Pour rings are autoclavable hydrophilic polypropylene
The updated design includes an improved ergonomic bottle shape and dedicated accessories for better handling, sample identification, and ease of use. The GL 45 bottles have a defined labelling area and an integrated retrace code for full traceability.