Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, PureAffin® R56™ for infiltration and embedding
Supplier: CANCER DIAGNOSTICS, INC.
PAR56J EEPAR56
10810-020CS 1672.02 CAD
10810-020 10810-022
Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, PureAffin® R56™ for infiltration and embedding
Paraffin wax blended with synthetic polymers
Infiltration and Embedding Paraffin. A blend of fully refined paraffin wax and select additives. A minimal compression histology wax (56-58°C)
  • Resealable stand-up bag
  • Also available in wide-mouth Plastic Jugs
Using Surgipath® Formula, try PureAffin® R56
CAS Number: 8002-74-2
IVDLogo