Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, PureAffin® 2™ for infiltration and embedding

Supplier: CANCER DIAGNOSTICS, INC.

PARA2
10810-014CS 770.9 CAD
10810-014
Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, PureAffin® 2™ for infiltration and embedding
Paraffin wax blended with synthetic polymers

PureAffin is specially formulated to provide ideal infiltration with all types of tissue specimens.
Use for both processing and embedding or with PureAffin® 1 Infiltration Medium as part of a two step system. Compatible with all industry paraffins. Blended to provide thorough impregnation of tissues and facilitate excellent sectioning characteristics. Use with open or closed tissue processors. A compound of purified paraffin and additives.


  • Stable at elevated temperatures
  • Melting Point 56-58°C (133-136F)
  • Working Temperature 60°C (140F)
  • Water Bath temperature 35-44°C (95-111F)
  • Store Below 30°C (86F)

CAS Number: 8002-74-2
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR