Dextran sulfate sodium salt, Molecular Biology Grade, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Dextran sulfate sodium from Dextran, Dextran sulphate sodium from Dextran

265152-10GM 265152-50GM
CA80502-348EA 256.38 CAD
CA80502-348 CA80502-350
Dextran sulfate sodium salt, Molecular Biology Grade, Calbiochem®
Dextran sulphate sodium salt

A long chain polymer of sulfated glucose, containing 17 - 20% sulfur. Has been used as an anticoagulant and has been shown to inhibit the binding of HIV-1 to CD4⁺ T lymphocytes. Used to accelerate hybridization of DNA probes to immobilized nucleic acids.

Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Refrigerator
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR