Lysonase™ Bioprocessing Reagent, EMD Millipore
Supplier: MilliporeSigma
71230-3 71230-4
CA82601-952EA 4385.89 CAD
CA82601-952 CA82601-954 CA82601-956
Lysonase™ Bioprocessing Reagent, EMD Millipore
Protein Extraction Protein Extraction Reagents
Lysonase™ bioprocessing reagent is an optimized, ready-to-use blend of rLysozyme™ solution and benzonase nuclease.
  • Increased protein extraction efficiency
  • Facilitate downstream processing of protein extracts
rLysozyme solution contains a highly purified and stabilized recombinant lysozyme with specific activity 250 times greater than that of chicken egg white lysozyme. Benzonase nuclease is a genetically engineered nonspecific endonuclease that degrades all forms of DNA and RNA (single stranded, double stranded, circular, linear), reducing extract viscosity and increasing protein yield.
For efficient protein extraction with BugBuster® protein extraction reagent, use 10 µl Lysonase per 1 g cell paste. For efficient protein extraction with PopCulture® reagent, add 2 µl Lysonase per 1 ml culture.