Paraffin wax blended with synthetic polymers, Solidification point: 50-54°C, Epredia™ Histoplast Extra

Supplier: RICHARD-ALLAN SCIENTIFIC CA

8332K 8332 8332
CA89212-368CS 367.54 CAD
10129-476 CA89212-368 89212-368
Paraffin wax blended with synthetic polymers, Solidification point: 50-54°C, Epredia™ Histoplast Extra
Paraffin wax blended with synthetic polymers

Meet the demanding and versatile needs of the laboratory with Epredia™ Histoplast Paraffin.


  • Made of paraffin and plastic polymers
  • Delivers excellent processing and yields thin, wrinkle–free sections
  • Also available prepackaged with Epredia™ Excelsior™ AS and Revos™ tissue processor waste wax containers for ease of use
  • Histoplast PE is good for general infiltration and embedding procedures (melting point 56°C)
  • Histoplast IM utilizes DMSO to help infiltrate larger tissues samples (melting point 56°C)
  • Histoplast LP has a lower melting point 50-54°C for small and friable tissue samples

Storage Temperature: Refrigerator
CAS Number: 8002-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR