Bismarck brown Y >45% (dye content), solid, certified

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: C.I. 21000, 1,3-Bis(2,3-diaminophenylazo)benzene dihydrochloride, Bismarck Brown Y (G), Basic Brown 1, Vesuvine, Phenylene Brown, Leather Brown, Basic Brown G, GX, GXP, Manchester Brown, 4,4'-[1,3-Phenylenebis(azo)]bisbenzene-1,3-diamine dihydrochloride

22887-100G 22887-25G
CH22887-100GEA 110.93 CAD
CH22887-100G CH22887-25G
Bismarck brown Y >45% (dye content), solid, certified
Bismarck Brown Y
Formula: C₁₈H₁₈N8·2HCl
MW: 419.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012977
CAS Number: 10114-58-6
Merck Index: 12,01294

Order Now

Specification Test Results

Infrared Spectrum Conforms to Structure
UV Ratios (in Ethanol + 5 mL 1N Hydrochloric Acid) λmax 457 nm
Dye Content ≥45%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR