ZymoBroth™ Media, Zymo Research Corporation
Supplier: Zymo Research
M3015-100 M3015-500
77001-970EA 60.44 CAD
77001-970 77001-972
ZymoBroth™ Media, Zymo Research Corporation
Media for Microbiology Prepared Media for Microbiology
Growth media for the preparation of highly competent E. coli cells.
  • Formulated growth mediua for making highly competent E. coli for DNA transformation
  • Choice growth media for difficult-to-transform E. coli strains
Compared to classic SOB growth medium, ZymoBroth increases transformation efficiency, typically on the order of 5 - 100 fold (depending on the E. coli strain). ZB media has been tested on a wide range of E. coli strains. ZB medium stimulates the transformation efficiency of all E. coli strains tested, including K12 derivatives (such as JM109, HB101 and more) and B strain derivatives (such as BL21 and more).