Organic Reference Standards, Medical Cannabis Terpenes Standards, Restek
Supplier: RESTEK CORP
34095
34096
76124-774EA
407.58
CAD
76124-774
76124-776
Organic Reference Standards, Medical Cannabis Terpenes Standards, Restek
Standards
Organic Standards
These standards are formulated to contain the major terpenes analyzed by medical cannabis labs.
- Comprehensive blends minimize time spent preparing standards for marijuana analysis
- High concentration provides value and maximize flexibility
The standards are prepared in two separate multicomponent mixes to maximize stability.