Endosafe® PTS™ Endotoxin Tests, Charles River
Supplier: CHARLES RIVER ENDOSAFE MS
PTS201 PTS-2005
MSPP-PTS201EA 910.77 CAD
MSPP-PTS201 MSPP-PTS2005
Endosafe® PTS™ Endotoxin Tests, Charles River
Endosafe®- PTS Endotoxin test utilizes existing FDA-licensed LAL formulations. The PTS provides quantitative LAL test results in approximately 15 minutes.
  • Reference package insert for storage requirements
  • Inhibition/enhancement screening
  • Rapid Micro Methods – Glucan
  • Acceptable temperature: 37 ±1 °C
  • Sample requirement: 25 μl sample per channel
Rapid micro methods Gram ID: Archived parameters for gram-negative, gram positive and yeast/mold isolate identification.