SVL® Solid Caps, With PTFE Protected Seals, Chemglass
Supplier: Chemglass
CG-188-01 CG-188-02 CG-188-03
80061-574EA 15.95 CAD
80061-574 80061-576 80061-578
SVL® Solid Caps, With PTFE Protected Seals, Chemglass
Closures Test/Sample Tube Closures
Solid SVL® caps with PTFE faced liner.
  • Cap and liner are autoclavable upto 121 °C
These caps are compatible for use with CG-187 screw threads.