WST-1 Cell Proliferation Assay Kit, Cayman Chemical
Supplier: Cayman Chemical Company
10008883-1
10008883-5
75816-714EA
127.92
CAD
75816-714
75816-716
WST-1 Cell Proliferation Assay Kit, Cayman Chemical
Assays
Cellular Assays
Used for detection of viable cells.
- A colorimetric, non-radioactive assay for assessing cell viability and proliferation
- Tetrazolium salt, WST-1, is reduced to a soluble purple formazan by trans-plasma membrane electron transport from NADH via an electron mediator
- Detect 1000 to 200000 cells per well
- Rapid assay; get results in 2 to 4 hours
Cayman’s WST-1 Proliferation Assay is based on the reduction of tetrazolium salt WST-1 to a soluble formazan dye by electron transport across the plasma membrane of dividing cells. This kit will also allow investigators to screen drug candidates involved in cell cycle regulation.