CHEMetrics® Filming Amines (aliphatic amine) Test Kits
Supplier: CHEMetrics
R-1000 K-1001
CXR-1000EA 216.67 CAD
CXR-1000 CXK-1001
CHEMetrics® Filming Amines (aliphatic amine) Test Kits
Environmental Test Kits
Filming amines are fed continuously into boiler feedwater to protect metal surfaces from corrosion caused by dissolved oxygen and carbon dioxide in condensate water. The amine forms a thin film on the surfaces that repels the potentially corrosive water.
CHEMetrics 3-minute procedure uses the standard methyl orange chemistry and features a unique extraction technique. The extraction eliminates several steps required in other procedures and provides increased sensitivity. The filming amine compound reacts with methyl orange to form a yellow-colored complex that is extracted into an immiscible organic solvent. Results are expressed in ppm (mg/L) octadecylamine.
Delivery information: Kit comes in a cardboard box and contains everything needed to perform, 20 tests: Refill, comparator, reaction tube with lid, tip breaking tool and instructions.