Orthophosphoric acid ≥75%, EMPROVE® EXPERT, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP
Danger

Synonyms: Phosphoric acid

1.00250.9950 1.00250.9010
CA1.00250.9010EA 582.04 CAD
CA1.00250.9010 EM1.00250.9950
Orthophosphoric acid ≥75%, EMPROVE® EXPERT, SAFC®
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 135 °C
Melting Pt: −17 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Assay (alkalimetric) 74.0- 76.0 %
Residual solvents (ICH Q3C) ... excluded by production process
Endotoxins ≤ 2.5 I.U./ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR