SATWipes® Polypropylene Wipes
Supplier: Contec
SWPP0004 SWCN0003 SWPP0003
76199-928CS 805.73 CAD
76199-928 76209-666 76209-740
SATWipes® Polypropylene Wipes
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Presaturated with IPA/DI water or 100% IPA.
  • Rolls of wipes conveniently dispensed through a canister
  • 0.2 micron (µm) filtered provides a safe, consistent and mild-cleaning solution
  • Eliminates the need for solvent bottles to reduce solvent usage and reduces solvent use by up to 35%
SATWIPES® Polypropylene wipes consists of a roll of (150) 6×9” saturated wipes dispensed through a center-pull canister with a pop-up dispenser. The wipes are made of soft and clean nonwoven polypropylene fabric. The solution is filtered to 0.2 µm and provides a safe,More Product Information